Would you like to log in again?
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2816-0100 BML-2816-0500
76002-334EA 609.89 USD InStock
76002-334 89156-278
SCREEN-WELL® Histaminergic Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Histaminergic and Melatonin Ligands Plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 680 ligands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty