Would you like to log in again?
SCREEN-WELL® Natural Product Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2865-0100 BML-2865-0500
76002-354EA 23228.07 USD InStock
76002-354 76002-356
SCREEN-WELL® Natural Product Library, Enzo Life Sciences
Cell Culture Supplements and Additives
Natural products are an unsurpassed source of chemical diversity and an ideal starting point for any screening program for pharmacologically active small molecules. Historically, natural products have been the most successful source of new drugs.
The Screen-Well® Natural Product library offers over 500 isolated natural compounds of known structure and includes many compounds from ethnopharmacology. Compounds are provided in solution in DMSO. Individual compounds or subsets can be resupplied in gram quantities.
Ordering information: Contains 502 compounds including compounds from the following classes of products: Terpenoids, Peptolides, Flavones, Coumarins, Alkaloids, Macrolides, Isoflavones, Synthetic derivates, and many more.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty