Would you like to log in again?
Matrix Metalloproteinase-19 (MMP-19) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK307-0001
76002-364EA 763.57 USD InStock
76002-364
Matrix Metalloproteinase-19 (MMP-19) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Assays Protein Assays
Designed to screen MMP-19 inhibitors using a quenched fluorogenic peptide.
The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-19, a potential therapeutic target. The compound NNGH is also included as a prototypic control inhibitor.
Ordering information: Contains 1 vial MMP-19 enzyme, 1 vial Substrate (OMNIMMP® RED), 1 vial 6’-TAMRA calibration standard, 1 vial control inhibitor (NNGH), 1 bottle (20 mL) assay buffer, 1 black 96-well microplate, Instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty