Would you like to log in again?
PROTEOSTAT® Protein Aggregation Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
51023-KP002 51023-KP050
89165-910EA 900.73 USD InStock
89165-910 76002-528
PROTEOSTAT® Protein Aggregation Assay, Enzo Life Sciences
Assays Protein Assays
PROTEOSTAT® Protein aggregation assay provides a simple, homogenous assay format for monitoring peptide and protein aggregation in solution
  • Validated for use with microplate or flow cytometry platform
  • Extensively benchmarked with IgG
  • Optimize buffers and excipients for protein formulation
  • Performs with a wide pH and ionic strength range
  • Quantitative detection of protein aggregates from visible to subvisible particles
The assay is capable of providing quantitative analysis of protein aggregation in a robust and high-throughput fashion (Z’ factor score >0.5). Lyophilized native and aggregated protein are provided as negative and positive controls for monitoring changes in protein aggregation status.
This is useful for defining optimal storage formulations for proteins, for screening of compounds that promote or inhibit protein aggregation and potentially for the sensitive measurement of molecular chaperone activity. The assay can be employed to streamline protein processing and formulation optimization procedures.
Delivery information: Kit includes PROTEOSTAT® detection reagent, PROTEOSTAT® positive control, PROTEOSTAT® negative control, 10X PROTEOSTAT® assay buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty