Would you like to log in again?
PROTEOSTAT® PDI Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
51024-KP050 51024-KP002
76002-530EA 287 USD InStock
76002-530 89165-912
PROTEOSTAT® PDI Assay Kit, Enzo Life Sciences
Assays Enzyme Assays
The PROTEOSTAT® PDI assay kit provides a simple, homogenous assay for screening modulators of PDI enzymatic activity.
The PROTEOSTAT® PDI assay kit is capable of providing a quantitative readout of PDI enzymatic activity in a robust and high-throughput fashion and can be applied to identification of PDI inhibitors from chemical libraries.
This is accomplished by monitoring PDI-catalyzed reduction of insulin in the presence of Dithiothreitol (DTT), resulting in the formation of insulin aggregates which then bind avidly to the red-emitting fluorogenic PROTEOSTAT® PDI detection dye.
Delivery information: Kit includes PROTEOSTAT® PDI detection reagent, PDI (human) (recombinant), Insulin (Lyophilized), Bacitracin (Inhibitor Control, Lyophilized), PBE buffer, Stop reagent, DTT (10 mM), Deionized water.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty