Would you like to log in again?
LEADING LIGHT® Wnt Cell Line Medium Pack, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-60003-0001
76002-536EA 491 USD InStock
76002-536
LEADING LIGHT® Wnt Cell Line Medium Pack, Enzo Life Sciences
Assays Cellular Assays
For use with the LEADING LIGHT® Wnt Reporter Cell Line.
  • Can be used as a supplement to the Wnt LEADING LIGHT® Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001)
  • When used in the Wnt LEADING LIGHT® Wnt Reporter Assay Starter Kit, there are many advantages of using this line of products:
  • True end-point detection system, high sensitivity (Wnt3a EC50 = 45.9 ng/ml)
  • Excellent reproducibility (Z'-factor of 0.74)
  • High signal-to-noise ratio without the need for Lithium Chloride to boost the signal
  • High-throughput screening applications (including 384-well microplates)
The LEADING LIGHT® Wnt Cell Line Medium Pack is intended for use with the LEADING LIGHT®Wnt Reporter Cell Line, which is the basis for a cell-based luciferase activity test suitable for a multi-well plate format, such as 96- or 384-well microplates
The LEADING LIGHT® Wnt Cell Line Medium Pack contains 60 mL, Cell Growth Medium Concentrate 4.5 mL, Cell Assay Medium Concentrate 10 mL, Cell Freezing Medium.
Note: The complete LEADING LIGHT® Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available.
The system contains an engineered 3T3 mouse fibroblast cell line, which expresses the firefly luciferase reporter gene under the control of Wnt-responsive promoters (TCF/LEF). The luciferase activity from the reporter gene in this cell line can be up-regulated in a dose-dependent manner upon the addition of exogenous Wnt protein/Wnt agonist or down-regulated by a further addition of a Wnt antagonist to the cell culture medium. This system can be used to elucidate the functions/activities of different Wnt-related ligands such as Wnt, DKK, etc. This system can also be used for screening small molecules and antibodies for their ability to act as Wnt inhibitors or Wnt agonists. The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty