Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NPC Mouse Monoclonal Antibody [clone: MAb414]

Supplier: Enzo Life Sciences
ABS105-0100
76002-572EA 526 USD InStock
76002-572
Anti-NPC Mouse Monoclonal Antibody [clone: MAb414]
Antibodies
Recommended Applications: ICC, IHC, IP, WB, Immunoelectron microscopy

Type: Primary
Antigen: NPC
Clonality: Monoclonal
Clone: MAb414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast, Xenopus, Vertebrates

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty