Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Mouse Monoclonal Antibody [clone: 11A50-B10]

Supplier: Enzo Life Sciences
ABS116-0200 ABS116-1000
76002-596EA 2800 USD InStock
76002-596 76002-598
Anti-APP Mouse Monoclonal Antibody [clone: 11A50-B10]
Antibodies
Recommended Applications: ELISA, IHC, WB

Species reactivity: Human, Mouse

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 11A50-B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty