Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRP Mouse Monoclonal Antibody [clone: 3F4]

Supplier: Enzo Life Sciences
ABS119-1000 ABS119-0500 ABS119-0200
76002-612EA 1770 USD InStock
76002-612 76002-610 76002-608
Anti-PRP Mouse Monoclonal Antibody [clone: 3F4]
Antibodies
Recommended Applications: ELISA, IHC, IP, WB

Species reactivity: Human, Hamster, Cat

Type: Primary
Antigen: PRP
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty