Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]

Supplier: Enzo Life Sciences
ABS122-0100
76002-622EA 797 USD InStock
76002-622
Anti-CD205 Rat Monoclonal Antibody [clone: NLDC-145]
Antibodies
Recommended Applications: Flow Cytometry, IF, IHC

Species reactivity: Mouse

Type: Primary
Antigen: CD205
Clonality: Monoclonal
Clone: NLDC-145
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty