Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40 Mouse Monoclonal Antibody (Biotin) [clone: mAb 89]

Supplier: Enzo Life Sciences
ABS146-0100
76002-682EA 593 USD InStock
76002-682
Anti-CD40 Mouse Monoclonal Antibody (Biotin) [clone: mAb 89]
Antibodies
Recommended Applications: Flow Cytometry, IHC (PS)

Species reactivity: Human

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: mAb 89
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty