Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMP3 Mouse Monoclonal Antibody (Biotin) [clone: 104G4]

Supplier: Enzo Life Sciences
ABS210-0100
76002-786EA 581 USD InStock
76002-786
Anti-LAMP3 Mouse Monoclonal Antibody (Biotin) [clone: 104G4]
Antibodies
Recommended Applications: Flow Cytometry, IHC (FS)

Species reactivity: Human, Sheep

Type: Primary
Antigen: LAMP3
Clonality: Monoclonal
Clone: 104G4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty