Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XBP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ABS255-0100
76002-862EA 547 USD InStock
76002-862
Anti-XBP1 Mouse Monoclonal Antibody
Antibodies
Recommended Applications: ELISA, ICC, IP, WB

Species reactivity: Human, Mouse

Type: Primary
Antigen: XBP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty