Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GST Mouse Monoclonal Antibody (Biotin) [clone: 8-326]

Supplier: Enzo Life Sciences
ABS280-0050
76002-910EA 426 USD InStock
76002-910
Anti-GST Mouse Monoclonal Antibody (Biotin) [clone: 8-326]
Antibodies
Recommended Applications: IP, WB

Species reactivity: Species independent

Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: 8-326
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat, Yeast, Rabbit, Insect, Hamster, E. coli, Chicken

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty