Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (DyLight™ 488)

Supplier: Enzo Life Sciences
ABS298-0100
76002-944EA 414 USD InStock
76002-944
Anti-IgG Rabbit Polyclonal Antibody (DyLight™ 488)
Antibodies
Recommended Applications: Flow Cytometry

Species reactivity: Species independent

Type: Isotype Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight™ 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty