Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KU55933 ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one

CHM150-0010 CHM150-0005
76003-098EA 165.74 USD InStock
76003-098 76003-096
KU55933 ≥99% (by HPLC)
KU55933
Potent and selective ATP-competitive inhibitor of ATM

KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer.


Formula: C₂₁H₁₇NO₃S₂
MW: 395,49 g/mol
MDL Number: MFCD08276985
CAS Number: 587871-26-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/16/73/45401673.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty