Would you like to log in again?
Cellular Senescence Activity Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT129-0120
76003-296EA 668 USD InStock
76003-296
Cellular Senescence Activity Assay, Enzo Life Sciences
Assays Cellular Assays
Highly reproducible SA-β-gal activity kit
  • Quantify SA-β-gal activity in a fluorescence plate reader
  • Easy-to-use protocol with results in less than 90 minutes
  • Fully compatible with all cell types
For the determination of cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. The 96-well Cellular senescence assay kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.
Ordering information: Contains 2X Cell lysis buffer, 2X Reaction buffer, SA-?-gal substrate (20X), Stop solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty