Would you like to log in again?
Human Recombinant Growth Hormone
Supplier: Enzo Life Sciences
PRT189-0010
76003-480EA 294.2 USD InStock
76003-480
Human Recombinant Growth Hormone
Proteins and Peptides
Produced in HEK cells. The human growth hormone (aa 27-217) is fused at the N-terminus to a FLAG®-tag followed by an 8 AA linker.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty