Would you like to log in again?
3 Small Flat Mop Head Frame, Berkshire
Supplier: Berkshire
BCR®
BCR.MOP3.1
76003-526EA 126.85 USD InStock
76003-526
3 Small Flat Mop Head Frame, Berkshire
Mops
Berkshire’s Flat Mop Head is light and easy to maneuver, even with wet wipers attached. The stainless steel springs or plastic eyes hold multiple mop wipers.
  • Clean, light, and durable
  • High quality anodized aluminum
  • Autoclavable (plastic eyes are not autoclavable)
  • Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® Mop Systems replicate the most effective way to clean a surface - wiping. Berkshire’s mop systems are easy to use, highly effective, and quick.
The system is very flexible, combining a wide range of alternative Mop Wipers designed for particular levels of cleanliness with a range of Flat Mop Heads.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty