Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Burette, PTFE Key Stopcock, Class A, Unserialized

Supplier: VWR
VWR®
115.452.03 115.452.02 115.452.04 115.452.01
76003-586CS 177.73 USD InStock
76003-586 76003-584 76003-588 76003-590
VWR® Burette, PTFE Key Stopcock, Class A, Unserialized
Burets
VWR® Burettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 heat-resistant glass.

  • All burettes have a bore size of 2 mm
  • Calibrated to deliver (TD, EX) at 20 °C
  • Calibrated to meet ASTM E 287 specifications
  • All burettes are calibrated according to Class A norms
  • A batch certificate is included

VWR® unserialized Class A burettes are used to deliver solution in precisely measured, variable volumes. They are used primarily for titration, to deliver one reactant until the precise endpoint is reached. Calibration and tolerances are in compliance to standards: ASTM 3287, ASTM E542, and ASTM E694.

Certifications: Burettes are supplied with Batch certificates.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty