Would you like to log in again?
VWR® Measuring Cylinder, Hexagonal Base, TD, with Spout, Class A, Unserialized
Supplier: VWR
VWR®
139.464.02A 139.464.05A 139.464.07A 139.464.06A 139.464.08A 139.464.04A 139.464.03A 139.464.01A
76003-696CS 467.46 USD InStock
76003-696 76003-686 76003-694 76003-684 76003-688 76003-692 76003-682 76003-690
VWR® Measuring Cylinder, Hexagonal Base, TD, with Spout, Class A, Unserialized
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Hexagonal base
  • Calibrated 'to Deliver'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certifications: Cylinders are supplied with Batch certificates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty