Would you like to log in again?
VWR® Volumetric Flask, Amber, Narrow Neck, Class B
Supplier: VWR
VWR®
134.431.03A 134.472.00BA 134.431.00A 134.422.01A 134.422.02A 134.422.03A 134.431.01A 134.472.02A 134.422.04A 134.472.07A 134.422.00A 134.431.02A 134.472.04A 134.431.08A 134.431.06A 134.472.00A 134.422.07A 134.431.04A 134.431.05A 134.472.05A 134.422.00BA 134.472.08A 134.422.06A 134.431.00BA 134.472.06A 134.422.08A 134.422.05A 134.472.01A 134.431.07A
76003-848CS 321.63 USD InStock
76003-848 76003-968 76003-846 76003-966 76003-648 76003-972 76003-752 76003-652 76003-850 76003-650 76003-970 76003-634 76003-976 76003-854 76003-974 76003-852 76004-066 76003-638 76003-858 76003-636 76003-978 76003-856 76003-642 76003-982 76003-640 76003-980 76003-844 76003-646 76003-644
VWR® Volumetric Flask, Amber, Narrow Neck, Class B
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty