Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Crossover Industrial Mixer, Caframo

Supplier: Caframo
CG6 A129 A128SET A118 1540 A016SET A356 1540 A354
76003-520EA 337.34 USD InStock
76003-520 76003-502 76003-778 76003-506 10834-024 CA10834-024 76003-518 76003-504 76003-516
Crossover Industrial Mixer, Caframo
Stirrers Overhead Stirrers
The Caframo 1540 combines user researched control features and incredible amounts of power creating a mixer unique to lab and industrial uses.

  • Brushless DC motor, fan cooled
  • 95% power transferred to shaft
  • Highest torque overhead mixer on market
  • Overload and overheat protection
  • Display; resistive touch screen
  • Operate with a gloved hand
  • Large (60×1100 mm) visibility of settings
  • Speed control on home screen
  • Bolt pattern allows mounting into custom systems

Resume on power up allows precise control by a PC or PLC. Plug in a USB drive for immediate data logging of date, speed, time and torque. Alarms, timers, PC programmability as well as 'mount and plug in' installation provide a transportable mixer from bench top to barrel.

Certifications: Conforms to applicable UL and CSA standards.

Ordering information: Clamp, stand, and impellers are available. Chuck, chuck shaft guard, chuck key, and power cord are included with the 1540 Mixer. Units are also supplied with a three-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty