Would you like to log in again?
SAMPL TUBE W/OUT CAP STERIL 5ML PK1000
Supplier: SIMPORT SCIENTIFIC INC.
Sample Tube With External Thread Sterile 5ml^ Sample Tube With External Thread, Sterile, Round bottom, Made of polypropylene, provides strength and clarity, non-toxic, autoclaved (121 degree C) in upright position with caps loosened, centrifuged up to 17,000g, Size: 12.5 x 92mm, Volume: 5ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPL TUBE W/OUT CAP STERIL 5ML PK1000 76003-808 T501-5TSP Pack of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty