Would you like to log in again?
VWR® Volumetric Flask, Clear Glass, Narrow Neck, Class B
Supplier: VWR
VWR®
134.472.05 134.472.02 134.472.00 134.472.04 134.472.01 134.472.08 134.472.07 134.472.06 134.472.00B
76003-920CS 264.92 USD InStock
76003-920 76003-930 76003-914 76003-924 76003-922 76003-918 76003-928 76003-916 76003-926
VWR® Volumetric Flask, Clear Glass, Narrow Neck, Class B
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty