Would you like to log in again?
VWR® Volumetric Flask, Amber, Wide Neck, Class B
Supplier: VWR
VWR®
131.431.05A 131.472.06A 131.431.03A 131.431.06A 131.431.01A 131.431.07A 131.472.05A 131.472.04A 131.431.10A 131.422.04A 131.422.03A 131.472.01A 131.472.09A 131.431.08A 131.472.08A 131.422.01A 131.422.02A 131.472.10A 131.472.07A 131.431.09A 131.422.06A 131.431.11A 131.422.11A 131.431.02A 131.422.10A 131.472.11A 131.422.08A 131.422.07A 131.431.04A 131.422.09A 131.422.05A
76003-804CS 770.98 USD InStock
76003-804 76003-868 76003-802 76003-988 76003-994 76003-796 76003-872 76003-794 76003-992 76003-870 76003-998 76003-876 76003-996 76003-654 76003-798 76003-874 76003-990 76003-792 76003-790 76004-000 76003-814 76003-878 76003-812 76003-862 76003-860 76003-800 76003-866 76003-986 76003-864 76003-984 76003-880
VWR® Volumetric Flask, Amber, Wide Neck, Class B
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class B Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty