Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

111554A 111574V 111564A 111554B 111574 111564B 111564 111564Y 111574R 111574G 111554 111554Y 111564G 111574N 111574Y 111564N 111554G 111574A 111564R 111574B 111554R 111554V 111564V 111554N
76004-078BG 26.31 USD InStock
76004-078 76004-122 76004-100 76004-110 76004-088 76004-102 76004-112 76004-104 76004-114 76004-106 76004-116 76004-092 76004-080 76004-094 76004-082 76004-096 76004-084 76004-120 76004-098 75845-358 76004-086 76004-090 76004-108 76004-118
Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
Tubes Microcentrifuge Tubes
The polypropylene (PP) tubes are resistant to most chemical reagents and can be autoclaved at 121 °C.

  • Now available in 8 attractive colors: amber, blue, green, natural, orange, red, violet, and yellow  
  • Efficient, automated production reduces cost and assures high quality
  • Tubes are made from USP Class VI medical grade polypropylene with excellent clarity
  • Supplied in resealable, tamper-evident, self standing bags
  • Tubes are graduated with frosted marking areas and piercable caps
  • Manufactured in a class 100000 cleanroom and certified to be free from RNase, DNase, DNA, and Pyrogens

Each tube is molded with precise graduation marks for accurate measurements. These microcentrifuge tubes are offered in resealable, tamper evident self-standing bags. 

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty