Would you like to log in again?
Locking Microcentrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
111582LK 111552LK 111572LK 111562LK
76004-124BG 38.3 USD InStock
76004-124 76004-126 76004-128 76004-130
Locking Microcentrifuge Tubes, Globe Scientific
Tubes Microcentrifuge Tubes
These tubes are engineered with a secure locking system that provides the security of a screw cap microtube with the simplicity of a snap cap tube. 
  • Maintain sample integrity with locking cap which prevents accidental cap opening and evaporation during contrifugation
  • Single-use tubes for laboratory procedures including centrifugation, boiling, incubation, freezing, and sample storage
  • Tubes are made from optimally clear USP Class VI medical grade polypropylene
  • Tubes feature graduations, writing area, piercable cap, are autoclavable, and are packaged in zip lock, re-closable bags
Manufactured in a class 100000 cleanroom and certified to be free from RNase and DNase, Human DNA, Pyrogens, PCR Inhibitors, and ATP.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty