Would you like to log in again?
Protein G Sepharose, Endotoxin-free, BioVision
Supplier: BioVision
AB286893-1ML AB286893-5ML AB286893-25ML
76004-500EA 712.47 USD InStock
76004-500 76004-502 76004-504
Protein G Sepharose, Endotoxin-free, BioVision
Chromatography Sorbents HPLC Sorbents
Endotoxin free Protein G Sepharose for High Specific Binding of IgG.
  • High flow rate
  • For high specific binding
  • Endotoxin-free
Protein G-Sepharose beads are prepared by covalently coupling recombinant Protein G to 6% cross-linked Sepharose beads under proprietary endotoxin-free condition
Purification of endotoxin-free monoclonal and polyclonal antibodies from culture media, serum, ascites fluid or hybridoma supernatants. Isolation of antibody/antigen complexes in immunoprecipitation experiments, since only the Fc region is involved in antibody binding and the Fab region is available for binding antigen.
BioVision Protein G (Cat. # 6510) is a genetically engineered protein containing three IgG-binding regions of native Protein G. The cell wall binding region, albumin binding region and other non-specific regions have been eliminated from the recombinant Protein G to ensure the maximum specific IgG binding. The coupling technique is optimized to give a higher binding capacity for IgG & minimum leaching of recombinant Protein G. The IgG binding capacity of Protein G-Sepharose is >20 mg of human or rabbit IgG per ml of wet beads. Protein G-Sepharose beads display high chemical & physical stability as well as high flow rate, hydrophilicity & high gel strength. It can be used for IgG purification and immunoprecipitation. Our Endotoxin-free Protein G-Sepharose also shows ability of reducing certain amount of endotoxin from serum or ascites samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty