Would you like to log in again?
96-Well Blocks, Zymo Research
Supplier: Zymo Research
P1002-2 P1001-10 P1001-2
76005-118EA 31.7 USD InStock
76005-118 76004-722 76005-116
96-Well Blocks, Zymo Research
Microplates Deep Well Microplates
2 mL deep-well block for processing and mixing of samples.
  • Conforms to laboratory standards
96-Well Block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2mL per round bottom well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty