Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Zymo-Spin™ Column/Filter Assemblies, Zymo Research
Supplier: Zymo Research
Zymo-Spin™
C1016-50 C1009-50 C1018-20 C1018-10 C1017-10 C1017-20 C1016-25 C1009-20
76004-198PK 60 USD InStock
76004-198 76005-170 76004-158 76004-160 76004-192 76005-168 76004-194 76004-196
Zymo-Spin™ Column/Filter Assemblies, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
  • For purifying DNA and or/RNA from high-volume sample inputs.
  • High-recovery.
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1.6μm pore size glass fiber filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty