Would you like to log in again?
PLATE DNA/RNA BINDING SILICON-A 600 UL
Supplier: Zymo Research
Silicon-A* Plate DNA/RNA binding^ Silicon-A* Plate, Format: DNA/RNA binding - up to 5 ug per well, Binding Capacity: 600ul per well, Elution Volume >/=30ul, Compatibility: Centrifuge, Matrix / Construction: Silica-based / polypropylene, Dimensions 19x 83x125 mm, Capacity: 600ul, size: 2 plates
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE DNA/RNA BINDING SILICON-A 600 UL 76005-208 C2001 Pack of 2 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty