Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Giant Polygon Magnetic Stirring Bars with Pivot Ring
Supplier: Avantor
VWR®
37122-6006 37122-6005
76006-382CS 1642.19 USD InStock
76006-382 76006-384
VWR® Giant Polygon Magnetic Stirring Bars with Pivot Ring
Stirrers Magnetic Stirring Bars
Designed for stirring large volumes in vessels such as drums and tanks
  • Marring of plastic containers is diminished
  • Multifaceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Ideal for use with the Cool Stir® Large Volume Stirrer
  • Capable of stirring volumes up to 190 L (50 gallons)
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
A molded pivot ring minimizes the contact area between the bar and the vessel, thus reducing friction and chattering.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty