Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Polygon Spinbar Magnetic Stirring Bars
Supplier: VWR
VWR®
37122-6060 37122-6020 37122-6070 37122-6025 37122-6035 37122-6045 37122-6040 37122-6012 37122-6050
76006-380CS 822.99 USD InStock
76006-380 76006-406 76006-402 76006-404 76006-388 76006-398 76006-378 76006-400 76006-386
VWR® Polygon Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty