Would you like to log in again?
Self-Igniting Economy Solder/Heat Tip Tool, Master Appliance, ORS Nasco
Supplier: ORS Nasco
467-UT-40SI
76006-438EA 81.45 USD InStock
76006-438
Self-Igniting Economy Solder/Heat Tip Tool, Master Appliance, ORS Nasco
Soldering Supplies Soldering Irons
Use this tool to pinpoint heat, remove surface mount components, repair vinyl, shrink heat shrinkable tubing, solder and desolder, spot dry, strip sulation with a hot knife, and more.
  • Convenient slide switch ignition system for quick ignition
  • New compact size, lightweight and completely portable
  • Rugged, easy-to-hold plastic body
  • Patented gas catalyst reaction system
  • Convenient On/Off gas control and adjustable temperature control
  • Durable tank has built-in fuel filter
  • Copper-based chisel tips provide long life and high thermal conductivity, excellent with lead-free solder
Unit cludes solder tip, heat tip, protective cap, iron, soldering and hot air tips.
1.3 hour run time. Butane gas fueled. Operating temerature range is 840 to 1,200 °F. 20 to 80W power output.
Certifications: UL Listed, TUV/GS; UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty