Would you like to log in again?
TUBE BRUSH SS WHITE 7.90IN F/FLEX ROD
Supplier: REMCO PRODUCTS
Vikan* Tube brush 7.90in^ Vikan* Tube brush for Flex rod, Material: Stainless Steel, Color: White, for cleaning pipes and other small areas, best used with flexible handles, Bristle Material: Polyester, Bristle: Medium, Autoclavable To: 250 F/121 C, Width: 0.40in, Height: 0.40in, Length: 7.90in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE BRUSH SS WHITE 7.90IN F/FLEX ROD 76006-800 53545 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty