Would you like to log in again?
TUBE BRUSH SS PURPLE 19.40IN CLEAN PIPE
Supplier: REMCO PRODUCTS
Vikan* Tube brush 19.40in^ Vikan* Tube brush, Material: Stainless Steel, Color: Purple, used to clean pipe, tab outlet & narrow space between machine part, Block Material: Polypropylene, Bristle Material: Polyester,: Medium, Bristle Length: 1.06in, Width: 2.50in, Height: 2.50in, Length: 19.40in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE BRUSH SS PURPLE 19.40IN CLEAN PIPE 76006-818 53708 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty