Would you like to log in again?
PADDLE SCRAPER BLADE STIFF PE RED 8.75IN
Supplier: REMCO PRODUCTS
Paddle Scraper Red^ Paddle Scraper, Stiff, Color: Red, scraping blade is used to remove remaining food stuff from containers prior to the cleaning procedure, Its rigid construction is useful when firm pressure needs to be applied into deep containers, Requires a European-thread handle
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PADDLE SCRAPER BLADE STIFF PE RED 8.75IN 76006-876 70114 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty