Would you like to log in again?
Vikan® Flexible Paddle Scraper, Remco
Supplier: Remco
Vikan®
70134
76006-884EA 19.52 USD InStock
76006-884
Vikan® Flexible Paddle Scraper, Remco
Scrapers
This scraping blade is used to remove remaining food stuff from containers prior to cleaning procedures.
  • Polyethylene construction
  • Euro (hygienic) thread type
  • Length: 8.75"
It is flexible and is used as a spatula for large or medium sized containers. Requires a European-thread handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty