Would you like to log in again?
BRUSH TURKS HEAD VIKAN PP GREEN 4.40IN
Supplier: REMCO PRODUCTS
Vikan* Turks Head Brush Soft Green^ Vikan* Turks Head Brush, Color: Green, Block Material: Polypropylene, soft bristle make it useful to clean area like small tanks, containers & for blending & fit well into meat mincer barrel, Require a European-thread handle & may be attached to any length handle
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BRUSH TURKS HEAD VIKAN PP GREEN 4.40IN 76006-888 70352 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty