Would you like to log in again?
Vikan® Blade Cleaning Brush with Hand Guard, Remco Products
Supplier: Remco
41845
76007-400EA 27.78 USD InStock
76007-400
Vikan® Blade Cleaning Brush with Hand Guard, Remco Products
Brushes
This brush is perfect for cleaning tools with exposed blades that could cause injury to the user.
  • Polyester bristles, polypropylene block
  • Dishwasher safe
  • Autoclavable up to 250°F
The narrow single row of bristles fits perfectly between the blades of any bowl cutter. The hand guard protects the user's hand from being cut during the cleaning process. Using this brush allows the user to clean the machine without disassembly.
Width: 5.5''
Height: 5.5''
Certifications: FDA compliant raw material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty