Would you like to log in again?
NAIL BRUSH PP PURPLE 4.90X1.90X1.50 IN
Supplier: REMCO PRODUCTS
Vikan* NailBrush Purple^ Vikan* NailBrush, small, Dimension: 4.90x1.90x1.50 in, color: Purple, Block Material: Polypropylene, Bristle Material: Polyester, Bristle Stiffness: Stiff, with FDA Compliant Material, Working Temp: -22 deg F/-30 deg C to 176 deg F/80 deg C, Autoclavable To: 250 deg F
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NAIL BRUSH PP PURPLE 4.90X1.90X1.50 IN 76007-786 64408 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty