Would you like to log in again?
SCOOP HAND POLYPROPYLENE GR LARGE/82OZ
Supplier: REMCO PRODUCTS
Hand Scoop Green^ Hand Scoop, Large, Dimension: 15.1x5.9x3.3 in, Color: Green, Material: Polypropylene, is great for handling bulk materials. Its solid construction makes it extremely durable. It has no seams or cracks to help prevent bacterial growth, 250 deg F/121 deg C, capacity: 82 oz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SCOOP HAND POLYPROPYLENE GR LARGE/82OZ 76007-792 65002 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty