Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Mega Dippers, Remco Products
Supplier: Remco
65526 65524 65522 65523 65525
76007-822EA 20.24 USD InStock
76007-822 76007-820 76007-824 76007-818 76007-816
Mega Dippers, Remco Products
Scoops
This fully color-coded dipper version of the mega scoop is great for handling bulk materials
Its solid construction makes it extremely durable. Polypropylene material.
W×H: 9 × 7"
Certifications: FDA compliant raw material.
Ordering information: A european-thread handle is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty