Would you like to log in again?
Vikan® Tank Brushes, Medium, Remco Products
Supplier: Remco
Vikan®
70394 70396 70393 70392 70395
76007-942EA 39.35 USD InStock
76007-942 76007-946 76007-944 76007-948 76007-950
Vikan® Tank Brushes, Medium, Remco Products
Brushes
Ideal for cleaning tanks and vats, and is also suitable for cleaning drain gulleys.
  • Block material is polypropylene; bristle material is polypropylene
  • Dishwasher safe
  • Autoclavable up to 250 °F
May be used with any European-thread handle from Vikan.
Certifications: FDA compliant raw material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty