Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vikan® Foam Blade Squeegee, 28", Remco Products

Supplier: Remco
77555
76008-392EA 40.1 USD InStock
76008-392
Vikan® Foam Blade Squeegee, 28", Remco Products
Mops
Excellent on all types of floors, including the grouting between tiles.

  • Made with FDA compliant materials
  • Autoclavable at 121 °C (250 °F)

The foam rubber blades on this squeegee make it the most effective model in the Vikan® range. It is excellent on all types of floors, as the soft rubber reaches into and cleans all spaces on the floor, such as grouting between tiles. The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical option. The foam blade squeegee may be used with any European-thread handle.

Accessories information: Replacement squeegee blades available, see 76008-214 or 76008-216.

Delivery information: Dropshipping available.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty