Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Round Bottom Flask, 3 Necks, Angled
Supplier: VWR
VWR®
060.402.28 060.402.27 060.402.36 060.402.25 060.402.37A 060.402.30A 060.402.29 060.402.21 060.402.30 060.402.17 060.402.20 060.402.24A 060.402.23 060.402.32 060.402.26 060.402.24
76012-820CS 208.82 USD InStock
76012-820 76008-630 76008-640 76012-812 76008-632 76008-642 76012-814 76008-634 76012-816 76008-644 76008-636 76008-646 76012-818 76008-638 76008-648 76008-628
VWR® Round Bottom Flask, 3 Necks, Angled
Flasks Round Bottom Flasks
Durable round bottom flasks with 3 necks at an angle.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty