Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Round Bottom Flask, 3 Necks, Parallel
Supplier: VWR
VWR®
061.402.19A 061.402.07A 061.402.19 061.402.12 061.402.01 061.402.07 061.402.16A 061.402.22 061.402.09 061.402.06 061.402.15 061.402.11 061.402.16 061.402.10A 061.402.18 061.402.13 061.402.13A 061.402.10 061.402.14 061.402.24
76008-670CS 159.36 USD InStock
76008-670 76008-680 76008-650 76008-672 76008-682 76008-660 76008-652 76008-674 76008-684 76008-662 76008-654 76008-676 76008-686 76008-664 76008-656 76008-678 76008-666 76008-688 76008-658 76008-668
VWR® Round Bottom Flask, 3 Necks, Parallel
Flasks Round Bottom Flasks
Durable round bottom flasks with 3 parallel necks.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty