Would you like to log in again?
VWR® Round Bottom Flask, 2 Necks, Angled
Supplier: VWR
VWR®
059.402.34 059.402.19 059.402.10 059.402.24 059.402.31 059.402.27 059.402.25 059.402.18 059.402.23 059.402.21
76012-790CS 135.08 USD InStock
76012-790 76012-792 76012-794 76012-782 76012-796 76012-784 76012-798 76012-786 76012-800 76012-788
VWR® Round Bottom Flask, 2 Necks, Angled
Flasks Round Bottom Flasks
Durable round bottom flasks with 2 necks at an angle.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 3.3 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty