Would you like to log in again?
VWR® 3D Cell Culture Dishes and Plates
Supplier: VWR
VWR®
TDD032035 TDD032060 TDD032070 TDP032024 TDP032006 TDP032012
76012-958CS 418.74 USD InStock
76012-958 76012-960 76012-952 76012-962 76012-954 76012-956
VWR® 3D Cell Culture Dishes and Plates
Culture Dishes Cell Culture Dishes
The 3D Scaffold for cell culture is able to simulate the three-dimensional structure of mamalian cells
  • 3D Scaffold is made from virgin polystyrene with a wire diameter of 500 µm
  • Scaffold spacing is a highly consistent 260 µm
  • Structured with 3-dimensional channels, offering extremely high connectivity to facilitate nutrient transmission
  • Consistency of metabolic activity and the accuracy of culture results in 3D cell culture
  • Cytokine and growth factor resistant
  • Easy cell secretion collection
Open pores with high connectivity, facilitating nutrient absorption and metabolism. 3D Scaffolds undergo strict integrity testing, and feature a larger surface area than regular cell culture products. Sterilized by electron beam irradiation. DNase/RNase-free and non-pyrogenic.
It provides an ideal environment for the interaction between cells, and improves the overall cell culture yield.
Certifications: ISO13485:2003 and ISO9001:2008.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty